missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CHIR-99021, 95%

Catalog No. AC468131000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468131000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468131000 Supplier Thermo Scientific™ Supplier No. 468131000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material CHIR-99021
CAS 252917-06-9
Synonym 6-[[2-[[4-(2,4-Dichlorophenyl)-5-(5-methyl-1 H-imidazol-2-yl)-2-pyrimidinyl]amino]ethyl]amino]-3-pyridinecarbonitrile
Quantity 100 mg
Molecular Formula C22 H18 Cl2 N8
Formula Weight 465.34
Physical Form Solid
Product Type Reagent
Purity 95%