missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ GSK-3 Inhibitor XVI

Catalog No. 3615595MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3615595MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3615595MG Supplier MilliporeSigma™ Supplier No. 3615595MG

GSK-3 Inhibitor XVI - CAS 252917-06-9, is a cell-permeable, potent, ATP-competitive, and highly selective GSK-3 inhibitor (IC50 = 10 and 6.7 nM against GSK-3α and GSK-3β, respectively).

Packaged Under Inert Gas

Specifications

Color Light Beige
Description 97% by HPLC
CAS 252917-06-9
Molecular Formula C22H18Cl2N8
Formula Weight 465.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg