missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ GSK-3 Inhibitor XVI, CHIR99021

Catalog No. 3615715MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3615715MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3615715MG Supplier MilliporeSigma™ Supplier No. 3615715MG

InSolution™ GSK-3 Inhibitor XVI, CHIR99021, CAS 252917-06-9, is a 25 mM solution in DMSO. A cell-permeable, ATP-competitive inhibitor of GSK-3 (IC50 = 10 & 6.7 nM for GSK-3α and GSK-3β, respectively).

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach

Specifications

Formulation A 25 mM (5mg/430μL) solution of GSK-3 Inhibitor XVI, CHIR99021 in DMSO.
Description 95% by HPLC
Target Proteins GSK-3 α and β
CAS 252917-06-9
Formula Weight 465.3g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg