CAS RN 21306-56-9
IUPAC Name:
{[(2,6-dimethylphenyl)carbamoyl]methyl}triethylazanium
Synonyms:
{[(2,6-dimethylphenyl)carbamoyl]methyl}triethylazanium
Molecular Weight (g/mol):
263.4
Molecular Formula:
C16H27N2O
InChi Key:
PYEBKOFMWAMBFV-UHFFFAOYSA-O
SMILES:
CC[N+](CC)(CC)CC(=O)NC1=C(C)C=CC=C1C
1
–
3
of
3
results
2
MilliporeSigma™ Calbiochem™ QX-314
A quaternary lidocaine derivative, permanently charged, and lipophobic.
3
Lidocaine N-ethyl bromide, 99+%, Thermo Scientific Chemicals
CAS: 21306-56-9 Molecular Formula: C16H27BrN2O Molecular Weight (g/mol): 343.309 MDL Number: MFCD00083182 InChI Key: DLHMKHREUTXMCH-UHFFFAOYSA-N Synonym: lidocaine n-ethyl bromide,qx 314 bromide,lidocaine n-ethyl bromide quaternary salt,n-2,6-dimethylphenylcarbamoylmethyl triethylammonium bromide,2,6-dimethylphenyl carbamoyl methyl triethylazanium bromide,d0k6yk,qx-314 bromide,qx-314.br,n-ethyllidocaine bromide PubChem CID: 9884487 IUPAC Name: [2-(2,6-dimethylanilino)-2-oxoethyl]-triethylazanium;bromide SMILES: CC[N+](CC)(CC)CC(=O)NC1=C(C=CC=C1C)C.[Br-]