missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine N-ethyl bromide, MilliporeSigma™ Supelco™

Catalog No. 111013538
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013538 50 mg
111013537 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013538 Supplier Merck Emd Millipore Supplier No. L578350MG
Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 21306-56-9
Physical Form Neat
Linear Formula C16H27 BrN2 O
MDL Number MFCD00083182
Synonym N-(2,6-Dimethylphenylcarbamoylmethyl)triethylammonium bromide; QX-314
Type Analytical Standard
Industry Type Drug Analysis
Quantity 50 mg
Formula Weight 343.30
Grade Analytical Standard