missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ QX-314

Catalog No. 552233100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
552233100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 552233100MG Supplier MilliporeSigma™ Supplier No. 552233100MG

A quaternary lidocaine derivative, permanently charged, and lipophobic.

A quaternary lidocaine derivative, permanently charged, and lipophobic. A powerful blocker of voltage-sensitive Na+ conductance when applied intracellularly. Suppresses the generation of Na+-dependent spikes from inside the cell membrane, without affecting Ca2+ currents or glutamate-activated currents.

Specifications

Description 99% by elemental analysis
Target Proteins A powerful blocker of voltage-sensitive Na+ conductance
CAS 21306-56-9
Molecular Formula C16H27BrN2O
Formula Weight 343.3g/mol
Solubility H2O
Content And Storage Protect from light
Form Lyophilized
Quantity 100 mg