accessibility menu, dialog, popup

UserName

NaOHNHNONOOSOOO(Z)

CAS RN 1787-61-7

IUPAC Name: sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate
Synonyms: eriochrome black T
Molecular Weight (g/mol): 461.38
Molecular Formula: C20H12N3NaO7S
InChi Key: JHUJLRKQZAPSDP-GXTSIBQPSA-M
SMILES: [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (23)
  • (2)
Quantity 
  • (2)
  • (1)
  • (7)
  • (1)
  • (7)
  • (1)
  • (1)
  • (3)
  • (2)
Color 
  • (3)
  • (4)
  • (5)
  • (5)
  • (2)
  • (2)
Grade 
  • (8)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
pH 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (14)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (9)
  • (23)
  • (2)
  • (2)
  • (1)
  • (7)
  • (1)
  • (7)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (4)
  • (5)
  • (5)
  • (2)
  • (2)
  • (8)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)