missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome Black T (1:100 diluted with NaCl), TCI America™

Catalog No. E04875G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04875G Supplier TCI America Supplier No. E04875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analytical reagent used as a metal indicator for complexometric titrations.

Chemical Identifiers

CAS 1787-61-7
Molecular Formula C20H12N3NaO7S
Molecular Weight (g/mol) 461.38
MDL Number MFCD00003935
InChI Key JHUJLRKQZAPSDP-GXTSIBQPSA-M
Synonym Mordant Black 11
PubChem CID 87355429
IUPAC Name sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O

Specifications

Color Black to Brown
Quantity 5 g
Physical Form Crystal/Powder
Formula Weight 461.38
Grade Indicator
Chemical Name or Material Eriochrome Black T (1:100 diluted with NaCl)
RTECSNumber QK2197000
TSCA Yes