missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome™ Black T, ACS Grade, LabChem™

Catalog No. LC140358
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC140358 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC140358 Supplier LabChem Supplier No. LC140358

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS grade analytical indicator suitable for use as a chemical indicator.

Chemical Identifiers

CAS 1787-61-7
Molecular Formula C20H12N3NaO7S
Molecular Weight (g/mol) 461.38
MDL Number MFCD00003935
InChI Key JHUJLRKQZAPSDP-GXTSIBQPSA-M
PubChem CID 87355429
IUPAC Name sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O

Specifications

CAS Max % 100
Identification Passes Test
Packaging Amber Glass
Suitability Passes Test
Linear Formula C20H12N3O7NaS
Decomposition Information Nitrogen oxides; Sulfur compounds; Carbon monoxide; Carbon dioxide
Color Black
Solubility Information Soluble in water
Quantity 25 g
Physical Form Powder
Formula Weight 461.38
Grade ACS
Chemical Name or Material Eriochrome™ Black T
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 217-250-3
Recommended Storage Room Temperature