missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Black T Indicator, 1% (w/w) in Sodium Chloride, Ricca Chemical
Specifications
Specifications
| Concentration | 1% (w/w) |
| Packaging | White Poly Wide Mouth Bottle |
| Solvent or Matrix | Sodium Chloride |
| Color | Gray |
| pH | 7 |
| Quantity | 100 g |
| Physical Form | Powder |
| Percent Purity | 1% w/w |
| Grade | Indicator |
| Chemical Name or Material | Eriochrome Black T Indicator |