Learn More
Thermo Scientific Chemicals Eriochrome Black T
Description
Eriochrome black T is used as an indicator in analytical chemistry. It is also used in the detection of rare earth metals. It finds application in dyeing and printing of wool fabric, dyeing of silk and nylon as well as in leather dyeing. It can be used in the spectrophotometric determination of diphenhydramine hydrochloride in pharmaceutical preparations. As a complexometric indicator, it is useful in the determination of hardness of water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Eriochrome black T is used as an indicator in analytical chemistry. It is also used in the detection of rare earth metals. It finds application in dyeing and printing of wool fabric, dyeing of silk and nylon as well as in leather dyeing. It can be used in the spectrophotometric determination of diphenhydramine hydrochloride in pharmaceutical preparations. As a complexometric indicator, it is useful in the determination of hardness of water.
Solubility
Soluble in water. Slightly soluble in alcohol and acetone.
Notes
Incompatible with strong oxidizing agents and strong reducing agents.
Specifications
Specifications
| Beilstein | 4121162 |
| Merck Index | 14,3667 |
| Solubility Information | Soluble in water. Slightly soluble in alcohol and acetone. |
| Quantity | 500 g |
| IUPAC Name | sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate |
| Formula Weight | 461.39 |
| Chemical Name or Material | Eriochrome Black T |
Safety and Handling
| Hazard Category | H319-H336-H500 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c |
| EINECSNumber | 217-250-3 |
| RTECSNumber | QK2197000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only