missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Black T, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Packaging | Pouch |
| Color | Black/Brown |
| Quantity | 25 g |
| Grade | ACS |
| Chemical Name or Material | Eriochrome Black T, Certified |
Safety and Handling
| ShelfLife | 60 months |