missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Eriochrome Black T Solution
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
Specifications
Specifications
| Identification | Pass Test |
| Packaging | EcoSafPak™ |
| Sensitivity | Pass Test |
| Density | 1.1 |
| Merck Index | 15, 3725 |
| Quantity | 500 mL |
| Formula Weight | 461.38 |
| Chemical Name or Material | Eriochrome Black T solution |
RUO – Research Use Only