CAS RN 16595-80-5
Levamisole Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Thermo Scientific Chemicals Levamisole hydrochloride, 99+%
CAS: 16595-80-5 Molecular Formula: C11H12N2S·ClH Molecular Weight (g/mol): 240.76 MDL Number: MFCD00012675 InChI Key: LAZPBGZRMVRFKY-HNCPQSOCSA-N Synonym: levamisole hydrochloride,levamisole hcl,ergamisol,tramisol,tramisole,decaris,--tetramisole hydrochloride,niratic hydrochloride,ascaridil,dekaris PubChem CID: 27944 ChEBI: CHEBI:6433 IUPAC Name: (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride SMILES: C1CSC2=NC(CN21)C3=CC=CC=C3.Cl
Levamisole Hydrochloride 99.0+%, TCI America™
CAS: 16595-80-5 Molecular Formula: C11H13ClN2S Molecular Weight (g/mol): 240.749 MDL Number: MFCD00012675 InChI Key: LAZPBGZRMVRFKY-HNCPQSOCSA-N Synonym: levamisole hydrochloride,levamisole hcl,ergamisol,tramisol,tramisole,decaris,--tetramisole hydrochloride,niratic hydrochloride,ascaridil,dekaris PubChem CID: 27944 ChEBI: CHEBI:6433 IUPAC Name: (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride SMILES: C1CSC2=NC(CN21)C3=CC=CC=C3.Cl
Levamisole Hydrochloride 99.0+%, TCI America™
CAS: 16595-80-5 Molecular Formula: C11H13ClN2S Molecular Weight (g/mol): 240.749 MDL Number: MFCD00012675 InChI Key: LAZPBGZRMVRFKY-HNCPQSOCSA-N Synonym: levamisole hydrochloride,levamisole hcl,ergamisol,tramisol,tramisole,decaris,--tetramisole hydrochloride,niratic hydrochloride,ascaridil,dekaris PubChem CID: 27944 ChEBI: CHEBI:6433 IUPAC Name: (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride SMILES: C1CSC2=NC(CN21)C3=CC=CC=C3.Cl
Levamisole hydrochloride, 99.62%, MP Biomedicals™
CAS: 16595-80-5 Molecular Formula: C11H13ClN2S Molecular Weight (g/mol): 240.749 InChI Key: LAZPBGZRMVRFKY-HNCPQSOCSA-N Synonym: levamisole hydrochloride,levamisole hcl,ergamisol,tramisol,tramisole,decaris,--tetramisole hydrochloride,niratic hydrochloride,ascaridil,dekaris PubChem CID: 27944 ChEBI: CHEBI:6433 IUPAC Name: (6S)-6-phenyl-2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazole;hydrochloride SMILES: C1CSC2=NC(CN21)C3=CC=CC=C3.Cl