missing translation for 'onlineSavingsMsg'
Learn More

Levamisole Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018832
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018832 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018832 Supplier Merck Emd Millipore Supplier No. PHR1798250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 16595-80-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C11H12N2 S · HCl
MDL Number MFCD00012675
Melting Point 228°C to 230°C
Synonym (-)-Tetramisole hydrochloride; (S)-(-)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride; L(-)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride; Levamisole hydrochloride
Type Certified Reference Material
Quantity 250 mg
Formula Weight 240.75
Grade Certified Reference Material