accessibility menu, dialog, popup

UserName

H3CSOO(R)OH(R)OHNHOClCl

CAS RN 15318-45-3

IUPAC Name: 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methanesulfonylphenyl)propan-2-yl]acetamide
Synonyms: thiamphenicol armai (TN) thiocymetin (TN) (+)-thiamphenicol 甲砜霉素
Molecular Weight (g/mol): 356.21
Molecular Formula: C12H15Cl2NO5S
InChi Key: OTVAEFIXJLOWRX-NXEZZACHSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)[C@@H](O)[C@@H](CO)NC(=O)C(Cl)Cl
Physical Form 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)