missing translation for 'onlineSavingsMsg'
Learn More

Thiamphenicol, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013225
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013225 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013225 Supplier Merck Emd Millipore Supplier No. 16715100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical structure: sulfonamide

Specifications

CAS 15318-45-3
Physical Form Neat
Linear Formula C12H15 Cl2 NO5 S
MDL Number MFCD00467983
Synonym D-threo-2,2-Dichloro-N-(beta-hydroxy-alpha-[hydroxymethyl]-4-[methylsulfonyl]phenethyl)acetamide
Solubility Soluble in ethanol
Type Analytical Standard
Quantity 100 mg
Formula Weight 356.22
Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Show More Show Less