accessibility menu, dialog, popup

UserName

CAS RN 147098-20-2

CaCH3CH3NNF(S)OH(R)OHOONCH3SCH3OOCH3H3CNNF(S)OH(R)OHOONH3CSCH3OO(E)(E)

CAS RN 147098-20-2

IUPAC Name: calcium bis((3R,5S,6E)-7-[4-(4-fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate)
Synonyms: calcium bis((3R,5S,6E)-7-[4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethanesulfonamido)pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate)
Molecular Weight (g/mol): 1001.14
Molecular Formula: C44H54CaF2N6O12S2
InChi Key: LALFOYNTGMUKGG-BGRFNVSISA-L
SMILES: [Ca++].CC(C)C1=NC(=NC(C2=CC=C(F)C=C2)=C1\C=C\[C@@H](O)C[C@@H](O)CC([O-])=O)N(C)S(C)(=O)=O.CC(C)C1=NC(=NC(C2=CC=C(F)C=C2)=C1\C=C\[C@@H](O)C[C@@H](O)CC([O-])=O)N(C)S(C)(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)