missing translation for 'onlineSavingsMsg'
Learn More

Rosuvastatin, calcium salt, 98%

Catalog No. AC456232500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456232500 250 mg
AC456230010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456232500 Supplier Thermo Scientific Chemicals Supplier No. 456232500
Only null left

CAS: 147098-20-2 | C22H28FN3O61/2Ca | 500.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 147098-20-2
Molecular Formula C22H28FN3O61/2Ca
Molecular Weight (g/mol) 500.58
InChI Key LQFSLFAVBQEFBH-LSMDDVBQSA-N
Synonym 3s,5r-rosuvastatin calcium salt, ent-rosuvastatin
PubChem CID 131673834
IUPAC Name calcium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid;molecular hydrogen
SMILES [HH].[HH].CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.[Ca]

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Formula Weight 500.58
Percent Purity 98%
Chemical Name or Material Rosuvastatin, calcium salt
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging the unborn child.
May cause harm to breast-fed children.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid contact during

RUO – Research Use Only