missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rosuvastatin Calcium 95.0+%, TCI America™
Specifications
Specifications
| Physical Form | Powder/Crystals |
| Color | White |
| Melting Point | 160°C |
| Quantity | 200 mg |
| Formula Weight | 1001.14 |
| Percent Purity | ≥95.0% (HPLC,N) |
| Chemical Name or Material | Rosuvastatin Calcium |
Safety and Handling
| RTECSNumber | MJ9675070 |
| TSCA | No |
| Recommended Storage | Freezer |