missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rosuvastatin Calcium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 147098-20-2 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C44H54 CaF2N6O12S2 |
| Synonym | Rosuvastatin calcium salt |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 1001.14 |
| Grade | Certified Reference Material |