CAS RN 145224-94-8
MES (Fine White Crystals), Fisher BioReagents
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N Synonym: 2-(4-Morpholino)ethane Sulfonic Acid,2-(N-Morpholino)ethanesulfonic acid hydrate PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES monohydrate, 99+%, for biochemistry
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N Synonym: 2-(N-Morpholino)ethanesulfonic acid monohydrate PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 7.5
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
MES, 1.0M buffer soln., pH 9.0, Thermo Scientific™
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
MES Monohydrate, ≥98.5% (Dry Basis), Ultrapure
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N Synonym: mes monohydrate,2-morpholinoethanesulfonic acid hydrate,mes hydrate,mes buffer hydrate,4-morpholineethanesulfonic acid monohydrate,4-morpholineethanesulfonic acid, monohydrate,2-4-morpholinyl ethanesulfonic acid hydrate,mes hydrate buffer,4-morpholineethanesulfonicacid, hydrate 1:1,mesmonohydrate PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 5.0
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES, 0.5M buffer soln., pH 6.5
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES, 0.2M buffer soln., pH 5.5
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES-buffered saline (5X), pH 6.5
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 6.5
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES monohydrate, 98%
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES, 0.2M buffer soln., pH 6.0
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES, 0.2M buffer soln., pH 6.0, low endotoxin
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 7.0
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
Thermo Scientific Chemicals MES, 0.5M buffer soln., pH 6.0
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.25 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate SMILES: O.OS(=O)(=O)CCN1CCOCC1
- 1
- 2