missing translation for 'onlineSavingsMsg'
Learn More

MES (Fine White Crystals), Fisher BioReagents

Catalog No. BP300100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP300100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP300100 Supplier Fisher BioReagents Supplier No. FLBP300 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Commonly used buffering agent

This biological buffer has a usable pH range of 5.5 to 6.7.

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym 2-(4-Morpholino)ethane Sulfonic Acid, 2-(N-Morpholino)ethanesulfonic acid hydrate
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Melting Point 300°C
Color White
pH 3.5 to 4.1
Physical Form Powder or Solid
Quantity 100 g
DNase DNase free
Identification Pass Test
Assay Percent Range ≥98 %
Linear Formula OCH2CH2NCH2CH2(CH2CH2SO3H)
Packaging Glass Bottle
Merck Index 15, 5972
Vapor Pressure Negligible
Formula Weight 213.2
Percent Purity ≥98%
Chemical Name or Material MES
Grade DNase- and RNase-Free
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. Wear personal protective equipment. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage RT