missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES-buffered saline (5X), pH 6.5

Catalog No. AAJ61979AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61979AP 500 mL
AAJ61979AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61979AP Supplier Thermo Scientific Chemicals Supplier No. J61979AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Concentration 5X
pH 6.5
Physical Form Liquid
Quantity 500 mL
Chemical Name or Material MES-buffered saline
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only