missing translation for 'onlineSavingsMsg'
Learn More

MES Monohydrate, ≥98.5% (Dry Basis), Ultrapure

Catalog No. AAJ18886A1
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ18886A1 1 kg Plastic bottle
AAJ1888622 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ18886A1 Supplier Thermo Fisher Scientific Supplier No. J18886A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym mes monohydrate, 2-morpholinoethanesulfonic acid hydrate, mes hydrate, mes buffer hydrate, 4-morpholineethanesulfonic acid monohydrate, 4-morpholineethanesulfonic acid, monohydrate, 2-4-morpholinyl ethanesulfonic acid hydrate, mes hydrate buffer, 4-morpholineethanesulfonicacid, hydrate 1:1, mesmonohydrate
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Melting Point ∼308°C (decomposition)
Packaging Plastic bottle
Linear Formula HO3S(CH2)2(C4H8NO)·H2O
Quantity 1 kg
Beilstein 141319
Formula Weight 213.25
Percent Purity ≥98.5%
Grade Ultrapure
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material MES Monohydrate
Show More Show Less
EINECSNumber 224-632-3
RTECSNumber QE3479500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only