missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES monohydrate, 98%

Catalog No. AAA1610436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1610436 500 g
AAA1610414 25 g
AAA1610422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1610436 Supplier Thermo Scientific Chemicals Supplier No. A1610436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

MES monohydrate is a biological buffer often referred to as Good’s buffer. It is useful for cell culture media and protein based buffer formulations to maintain a stable environment in solution. It is also used in the purification bioprocesses of antibodies, peptides, proteins, blood components and growth factors. It is a useful resolving agent for very small proteins on bis-tris gels. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES monohydrate is a biological buffer often referred to as Good’s buffer. It is useful for cell culture media and protein based buffer formulations to maintain a stable environment in solution. It is also used in the purification bioprocesses of antibodies, peptides, proteins, blood components and growth factors. It is a useful resolving agent for very small proteins on bis-tris gels. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00149409
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Melting Point ∽308°C (decomposition)
Quantity 500 g
Assay Percent Range 98%
Beilstein 141319
Merck Index 14,5902
Solubility Information Soluble in water.
Formula Weight 213.26 (195.24 Anhydrous)
Percent Purity 98%
Chemical Name or Material MES Monohydrate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
EINECSNumber 224-632-3
RTECSNumber QE3479500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only