accessibility menu, dialog, popup

UserName

CuO

CAS RN 1317-38-0

IUPAC Name: copper(2+) oxidandiide
Synonyms: copper(II) oxide
Molecular Weight (g/mol): 79.55
Molecular Formula: CuO
InChi Key: KKCXRELNMOYFLS-UHFFFAOYSA-N
SMILES: [O--].[Cu++]
Physical Form 
  • (1)
  • (26)
  • (5)
  • (5)
  • (5)
  • (4)
Quantity 
  • (4)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (4)
  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (10)
Percent Purity 
  • (4)
  • (1)
  • (4)
  • (2)
  • (5)
  • (5)
  • (3)
  • (5)
Grade 
  • (8)
  • (3)
  • (5)
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (10)
  • (2)
  • (10)
  • (9)
  • (25)
  • (1)
  • (26)
  • (5)
  • (5)
  • (5)
  • (4)
  • (4)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (4)
  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (10)
  • (4)
  • (1)
  • (4)
  • (2)
  • (5)
  • (5)
  • (3)
  • (5)
  • (8)
  • (3)
  • (5)
  • (2)
  • (5)