missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Oxide, Granular, For Elementary Analysis, MilliporeSigma™

Catalog No. M1027640250
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1027640250 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1027640250 Supplier Technidata Supplier No. 1.02764.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity granules designed for reliable performance in elementary analysis applications.

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
IUPAC Name copper(2+) oxidandiide
SMILES [O--].[Cu++]

Specifications

Physical Form Solid
Quantity 250 g
UN Number UN 3077
Formula Weight 79.55
Percent Purity ≥99%
Odor Odorless
Packaging Plastic bottle
Appearance Black solid
Chemical Name or Material Copper(II) oxide
Hazard Category H302: Harmful if swallowed.
H410: Very toxic to aquatic life with long lasting effects.