Learn More
Copper(II) oxide, ACS, 99.0% min
Description
Copper(II) oxide is used as an adsorbent and absorbent, pigment, antifouling agent, wood preservative, optical glass polishing agent, welding flux for bronze materials, agricultural chemical and abrasive. It is used in the preparation of intermediates, oxidizing/reducing agents, surface treating agents and process regulators. It finds use in the manufacture of rayon, phosphors and dry battery cells. It serves as a catalyst in certain chemical reactions of industrial importance. It is also employed in chemical sensors and superconducting materials. Hazardous materials like cyanides, dioxins and halogenated hydrocarbons can be disposed through oxidation with copper oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Water softener, metal cleaner, soap and synthetic detergent builder, general sequestering agent, in electrodeposition of metals
Specifications
Specifications
| Melting Point | 1326°C |
| Color | Black |
| Physical Form | Powder |
| Quantity | 1 kg |
| Odor | Odorless |
| Refractive Index | 2.63 |
| Appearance | Black powder |
| Assay Percent Range | 99.0% min. |
| Density | 6.3 |
| Solubility Information | Practically insoluble in water and alcohol. Soluble in dilute acids,alkali cyanides and ammonium carbonate solution. |
| Show More |
Safety and Handling
| Hazard Category | H400-H410-H302; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. |
| DOTInformation | DOT Class: 9, Packaging Group: III |
| EINECSNumber | 215-269-1 |
| RTECSNumber | GL7900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only