missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) Oxide Powder, Extra Pure, MilliporeSigma™
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 25 kg |
| Odor | Odorless |
| Packaging | Plastic drum |
| Appearance | Black solid |
| UN Number | UN 3077 |
| Formula Weight | 79.55. (molar mass) |
| Grade | Extra Pure |
| Chemical Name or Material | Copper(II) Oxide Powder |
Safety and Handling
| Hazard Category | H302: Harmful if swallowed. H410: Very toxic to aquatic life with long lasting effects. |
| DOTInformation | DOT Class: 9, Packaging Group: III |
| EINECSNumber | 215-269-1 |