CAS RN 13106-76-8
IUPAC Name:
diammonium dioxomolybdenumbis(olate)
Synonyms:
ammonium orthomolybdate
Molecular Weight (g/mol):
196.02
Molecular Formula:
H8MoN2O4
InChi Key:
APUPEJJSWDHEBO-UHFFFAOYSA-P
SMILES:
[NH4+].[NH4+].[O-][Mo]([O-])(=O)=O
1
–
12
of
12
results
1
Ammonium molybdate, 99.99%, (trace metal basis)
CAS: 13106-76-8 Molecular Formula: H8MoN2O4 Molecular Weight (g/mol): 196.01
2
Promotions Available
Ammonium molybdate, 99.99% (metals basis)
Ammonium molybdate, 99.99% (metals basis)
CAS: 13106-76-8 Molecular Formula: H8MoN2O4 MDL Number: MFCD00011596
6
7
Molybdenum AA Standard, 1000 ppm Mo in 3% NH4OH, Ricca Chemical
CAS: 13106-76-8
8
9
Molybdenum ICP Standard, 1000 ppm Mo in 3% NH4OH, Ricca Chemical
CAS: 13106-76-8
11
Molybdenum AA Standard, 1000 ppm Mo in 3% HNO3, Ricca Chemical
CAS: 13106-76-8
12
Molybdenum Standard For ICP/MS, SPEX CertiPrep™
CAS: 13106-76-8