missing translation for 'onlineSavingsMsg'
Learn More

SIC I Standard: 500 ppm Mo in H2O, Ricca Chemical

Catalog No. RSIC1X00500
Change view
Click to view available options
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
RSIC1X00500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSIC1X00500 Supplier Ricca Chemical Company Supplier No. RSIC1X00500N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Single Element ICP Standard

Specifications

Packaging Natural Poly Bottle
CAS 13106-76-8, 1336-21-6, 7732-18-5
CAS Min % 99.88, 0.1, 0.02
CAS Max % 99.88, 0.1, 0.02
Density 1.02
Molecular Formula H8MoN2O4
Solubility Information Miscible in water
InChI Key APUPEJJSWDHEBO-UHFFFAOYSA-P
Molecular Weight (g/mol) 196.02
Chemical Name or Material SIC I Standard: 500 ppm Mo in H2O
ShelfLife 12 months
Recommended Storage Ambient