missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum AA Standard, 1000 ppm Mo in 3% HNO3, Ricca Chemical

Catalog No. AMO1KN100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AMO1KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMO1KN100 Supplier Ricca Chemical Company Supplier No. AMO1KN100

missing translation for 'validMainframeMsg'

Specifications

Packaging Natural LDPE Bottle
CAS Min % 94.0, 4.22, 0.19
CAS Max % 97.0, 4.39, 0.2
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg Mo (1, 000ppm Mo)(NH4)2MoO4 in 3% HNO3
Chemical Name or Material Molybdenum AA Standard
Show More Show Less
ShelfLife 24 months