missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum ICP Standard, 1000 ppm Mo in 3% NH4OH, Ricca Chemical

Catalog No. PMO1KW100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PMO1KW100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PMO1KW100 Supplier Ricca Chemical Company Supplier No. PMO1KW100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13106-76-8 , 1336-21-6 , 7732-18-5

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Mo
CAS Min % 95.0, 2.85, 0.19
CAS Max % 99.0, 2.96, 0.2
Equivalence 1 mL = 1 mg Mo
Physical Form Liquid
Color Colorless to Green/Yellow
Boiling Point 100°C
Type Standard
Quantity 100 mL
Chemical Name or Material Molybdenum ICP Standard
Show More Show Less