Learn More
Ammonium molybdate, 99.99% (metals basis)
Description
It is used to prepare phosphomolybdates, to measure the amount of phophates, silicates, arsenates, lead in pigments, river water and sea water. It is also used as a negative stain in electron microscopy, in photogrphy, ceramics, paints. It is used in the production of molybdenum. It is used in the preparation of dehydrogenation and desulphurization catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Lump/Powder |
| Quantity | 100 g |
| Odor | Odorless |
| Assay Percent Range | 99.99% (Metals basis) |
| Solubility Information | Soluble in water,acid and alkali,but insoluble in alcohol. |
| Formula Weight | 196.02 |
| Chemical Name or Material | Ammonium molybdate |
Safety and Handling
| Hazard Category | H302-H500 |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 236-031-3 |
| RTECSNumber | QA4900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only