missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum ICP Standard, 1000 ppm Mo in 3% NH4OH, Ricca Chemical

Catalog No. PMO1KW500
Change view
Click to view available options
Package: Natural Poly; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PMO1KW500 Package: Natural Poly; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PMO1KW500 Supplier Ricca Chemical Company Supplier No. PMO1KW500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural Poly Bottle
CAS 13106-76-8, 1336-21-6, 7732-18-5
Concentration or Composition Notes 1,000ppm Mo
CAS Min % 95.0, 2.85, 0.19
CAS Max % 99.0, 2.96, 0.2
Equivalence 1 mL = 1 mg Mo
Chemical Name or Material Molybdenum ICP Standard