accessibility menu, dialog, popup

UserName

CAS RN 121-32-4

IUPAC Name: 3-ethoxy-4-hydroxybenzaldehyde
Synonyms: ethyl vanillin ethylvanillin 3-ethoxyprotocatechualdehyde protocatechuic aldehyde ethyl ether ethylprotocatechuic aldehyde vanirome
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: CBOQJANXLMLOSS-UHFFFAOYSA-N
SMILES: CCOC1=CC(C=O)=CC=C1O
Percent Purity 
  • (2)
  • (3)
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
Grade 
  • (5)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (5)
  • (2)
  • (6)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (5)
  • (2)
  • (3)
  • (3)