missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Vanillin, NF, 98-101%, Spectrum™ Chemical

Catalog No. 18602553
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
40 kg
Packaging:
Amber Glass Bottle
Drum
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602553 40 kg Drum
18602549 125 g Amber Glass Bottle
18602550 500 g Amber Glass Bottle
18602551 2.5 kg Poly Pail
18602552 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602553 Supplier Spectrum Chemical Mfg Cor Supplier No. ET11940KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ET119, 121-32-4, C9H10O3

Spectrum™ Chemical Ethyl Vanillin, NF is used as a flavorant. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 121-32-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00006944
InChI Key CBOQJANXLMLOSS-UHFFFAOYSA-N
IUPAC Name 3-ethoxy-4-hydroxybenzaldehyde
SMILES CCOC1=CC(C=O)=CC=C1O

Specifications

CAS Max % 100%
Loss on Drying 1%
Melting Range 76-78°C
Packaging Drum
Quantity 40 kg
Percent Purity 98 to 101%
Grade NF