missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vanillin, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012763
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012763 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012763 Supplier Merck Emd Millipore Supplier No. 75042100MG
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 121-32-4
Vapor Pressure <0.01 mmHg (25°C)
Flash Point 145°C - Closed Cup
Physical Form Neat
Linear Formula C2H5 OC6H3(OH)CHO
MDL Number MFCD00006944
Melting Point 74°C to 77°C
Synonym 3-Ethoxy-4-hydroxybenzaldehyde; Ethylvanillin; NSC 1803; NSC 67240
Type Analytical Standard
Quantity 100 mg
Formula Weight 166.17
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less