missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-4-hydroxybenzaldehyde, 98%

Catalog No. AAA1947836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1947836 500 g
AAA1947814 25 g
AAA1947822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1947836 Supplier Thermo Scientific Chemicals Supplier No. A1947836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121-32-4 | C9H10O3 | 166.18 g/mol

It plays an important role as an antioxidant and is also used as a flavoring agent, in perfumes, and in organic synthesis. It holds anti-angiogenic, anti-inflammatory and antinociceptive properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It plays an important role as an antioxidant and is also used as a flavoring agent, in perfumes, and in organic synthesis. It holds anti-angiogenic, anti-inflammatory and antinociceptive properties.

Solubility
Slightly soluble in water. Soluble in ethanol, ether, benzene.

Notes
Air & light sensitive. Protect from light. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 121-32-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00006944
InChI Key CBOQJANXLMLOSS-UHFFFAOYSA-N
Synonym ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal
PubChem CID 8467
ChEBI CHEBI:48408
IUPAC Name 3-ethoxy-4-hydroxybenzaldehyde
SMILES CCOC1=CC(C=O)=CC=C1O

Specifications

Melting Point 75°C to 79°C
Boiling Point 285°C
Flash Point 127°C (260°F)
Quantity 500 g
Beilstein 1073761
Sensitivity Air and light sensitive
Merck Index 14,3859
Solubility Information Slightly soluble in water. Soluble in ethanol,ether,benzene.
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 3-Ethoxy-4-hydroxybenzaldehyde
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 204-464-7
RTECSNumber CU6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only