accessibility menu, dialog, popup

UserName

H2NOHOCl

CAS RN 1134-47-0

IUPAC Name: 4-amino-3-(4-chlorophenyl)butanoic acid
Synonyms: baclofen nu baclo novartis brand of baclofen lioresal intrathecal medtronic brand of baclofen NuBaclo baclofen awd baclofen alphapharm brand baclofene baclofeno
Molecular Weight (g/mol): 213.66
Molecular Formula: C10H12ClNO2
InChi Key: KPYSYYIEGFHWSV-UHFFFAOYNA-N
SMILES: NCC(CC(O)=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)