missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baclofen 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 208°C |
| UN Number | 2811 |
| Quantity | 5 g |
| Formula Weight | 213.66 |
| Percent Purity | ≥98.0% (T) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Baclofen |
Safety and Handling
| RTECSNumber | MW5084200 |
| TSCA | No |