missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Baclofen, 99+%

Catalog No. AAJ6276803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6276803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6276803 Supplier Thermo Scientific Chemicals Supplier No. J6276803

CAS: 1134-47-0 | C10H12ClNO2 | 213.661 g/mol

Selective GABA-b receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1134-47-0
Molecular Formula C10H12ClNO2
Molecular Weight (g/mol) 213.661
MDL Number MFCD00055143
InChI Key KPYSYYIEGFHWSV-UHFFFAOYSA-N
Synonym baclofen, lioresal, baclon, 4-amino-3-4-chlorophenyl butanoic acid, dl-baclofen, kemstro, gabalon, baclofene, baclofeno, baclofenum
PubChem CID 2284
ChEBI CHEBI:2972
IUPAC Name 4-amino-3-(4-chlorophenyl)butanoic acid
SMILES C1=CC(=CC=C1C(CC(=O)O)CN)Cl

Specifications

Color White
UN Number UN2811
Merck Index 14,937
Solubility Information Soluble in dilute NaOH or dilute HCl; Soluble in water at approximately 4mg/ml at pH 7;6
Quantity 1 g
Formula Weight 213.66
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material (+/-)-Baclofen
Hazard Category H301-H315-H317-H319-H334-H335-H360
Hazard Statement GHS H Statement
H301-H334-H360-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 214-486-9
RTECSNumber MW5084200
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only