missing translation for 'onlineSavingsMsg'
Learn More

Baclofen Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018956
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018956 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018956 Supplier Merck Emd Millipore Supplier No. PHR16821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Baclofen is a structural analog of γ-aminobutyric acid (GABA) and an antispastic agent commonly used for the treatment of spasticity as a result of spinal cord or brain injury. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 1134-47-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C10H12 ClNO2
MDL Number MFCD00055143
Synonym (+/-)-Baclofen; (+/-)-beta-(Aminomethyl)-4-chlorobenzenepropanoic acid; Lioresal
Type Certified Reference Material
Quantity 1 g
Formula Weight 213.66
Grade Certified Reference Material