accessibility menu, dialog, popup

UserName

CAS RN 1119-97-7

IUPAC Name: trimethyl(tetradecyl)azanium bromide
Synonyms: tetradonium bromide|myrtrimonium bromide|alkyl* trimethyl ammonium bromide *(95%C14, 5%C16)
Molecular Weight (g/mol): 336.4
Molecular Formula: C17H38BrN
InChi Key: CXRFDZFCGOPDTD-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (9)
Percent Purity 
  • (3)
  • (1)
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
  • (4)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (3)
  • (6)
  • (1)
  • (9)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)