missing translation for 'onlineSavingsMsg'
Learn More

Myristyltrimethylammonium bromide, 99%

Catalog No. AC229375000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229375000 500 g Plastic bottle
AC229370050 5 g Glass bottle
AC229371000 100 g Plastic bottle
AC229370025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC229375000 Supplier Thermo Scientific Chemicals Supplier No. 229375000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1119-97-7 | C17H38BrN | 336.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 1119-97-7
Molecular Formula C17H38BrN
Molecular Weight (g/mol) 336.40
MDL Number MFCD00011770
InChI Key CXRFDZFCGOPDTD-UHFFFAOYSA-M
Synonym cetrimide, tetradecyltrimethylammonium bromide, myristyltrimethylammonium bromide, tetradonium bromide, n,n,n-trimethyltetradecan-1-aminium bromide, tetradecyl trimethyl ammonium bromide, myrtrimonium bromide, mytab, mitmab, morpan t
PubChem CID 14250
ChEBI CHEBI:3565
IUPAC Name trimethyl(tetradecyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 245°C to 250°C
Color White
pH 6 to 8 (1% aq. soln.)
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (Argentometry)
Packaging Plastic bottle
Linear Formula CH3(CH2)13N(CH3)3Br
Quantity 500 g
Merck Index 15,6422
Solubility Information Solubility in water: 100g/L (20°C)
Formula Weight 336.39
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Myristyltrimethylammonium bromide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 214-291-9
RTECSNumber BS5776000
TSCA TSCA

RUO – Research Use Only