missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Myristyltrimethylammonium bromide, ≥99.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111018569
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018569 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018569 Supplier Merck Emd Millipore Supplier No. 8720810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity reagent suitable for ion pair chromatography.

Chemical Identifiers

CAS 1119-97-7
Molecular Weight (g/mol) 336.39
MDL Number MFCD00011770
Synonym Tetradecyltrimethylammonium bromide; Trimethyl(tetradecyl)ammonium bromide

Specifications

Melting Point 245°C to 250°C
Physical Form Crystals
Quantity 10 g
Linear Formula CH3(CH2)13 N(Br)(CH3)3
Formula Weight 336.39
Percent Purity ≥99.0% (AT)
Grade Ion Pair Chromatography
Product Line LiChropur
Chemical Name or Material Myristyltrimethylammonium Bromide
RTECSNumber BS5776000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany