missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Dynamin Inhibitor II, MiTMAB

Catalog No. 324411500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
324411500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 324411500MG Supplier MilliporeSigma™ Supplier No. 324411500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Dynamin Inhibitor II, MiTMAB, also referenced under CAS 1119-97-7, controls the biological activity of Dynamin. This small molecule/inhibitor is primarily used for Membrane applications.

Packaged Under Inert Gas

Specifications

Color White
Description 99% by Titration
CAS 1119-97-7
Molecular Formula C17H38NBr
Formula Weight 336.4g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic
Form Solid
Quantity 500 mg