accessibility menu, dialog, popup

UserName

ClNH2(S)NH2NNH2OHOH

CAS RN 1119-34-2

IUPAC Name: hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
Synonyms: arginine hcl
Molecular Weight (g/mol): 210.66
Molecular Formula: C6H15ClN4O2
InChi Key: KWTQSFXGGICVPE-WCCKRBBISA-N
SMILES: [H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O
Molecular Weight (g/mol) 
  • (12)
  • (14)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (14)
  • (1)
Quantity 
  • (5)
  • (5)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (3)
Grade 
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (8)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (18)
  • (15)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (12)
  • (2)
  • (2)
  • (5)
  • (12)
  • (14)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (14)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (8)
  • (2)
  • (1)