missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine Hydrochloride (White Crystals or Crystalline Powder), Fisher BioReagents

Catalog No. BP372100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP372100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP372100 Supplier Fisher BioReagents Supplier No. BP372 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The hydrochloride salt of L-Arginine is suitable for use in tissue culture systems requiring additives and has many of the same applications as L-Arginine.

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
PubChem CID 66250
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Melting Point 226°C
Ammonium 0.02% max.
Boiling Point 235°C
Color White
Chloride 16.58 to 17.00%
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Identification Pass Test
Loss on Drying 0.2% max.
Assay Percent Range 98.5 to 101.0 %
Other Amino Acids Not Detected
Packaging Glass Bottle
Specific Rotation 22 to 23° (+ or -)
Merck Index 15, 770
State of Solution (%T) 98.0% min.
Alpha Vector Arginine Hydrochloride
Quantity 100 g
Formula Weight 210.58
Percent Purity 98.5 to 101.0%
Physical Form Solid
Chemical Name or Material L-Arginine Hydrochloride
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT